Learn More
AdipoGen 6BIO (25 mg)
SDP
Supplier:  AdipoGen AGCR10056M025
Encompass_Preferred
Chemical. CAS: 667463-62-9. Formula: C16H10BrN3O2. MW: 356.2. Phosphoinositide-dependent kinase 1 (PDK1) inhibitor. Potent, reversible and ATP-competitive glycogen synthase kinase-3alpha/beta (GSK-3alpha/beta) inhibitor. JAK/STAT3 signaling inhibitor. Apoptosis inducer. Potent antiproliferative agent in malignant lymphoid cell. Antimetastatic agent. Potent chemosensitizing agent fighting TRAIL resistant cancer cells.
Catalog No. 50-205-3814