Learn More
Medchemexpress LLC 6bK TFA | 1774353-12-6 | 99.7% | 871.94 | 10 MG
SDP
Catalog No. 5000344048
Encompass_Preferred
6bK TFA is a potent and selective inhibitor of insulin degrading enzyme (IDE) with an IC50 value of 50 nM. It increases circulating insulin in high-fat-fed mice and enhances glucose tolerance to oral glucose, notably to a greater extent in high-fat-fed mice. This product is suitable for type 2 diabetes research.
  • Potent and selective inhibitor of insulin degrading enzyme (IDE)
  • Increases circulating insulin in high-fat-fed mice
  • Enhances glucose tolerance to oral glucose
  • Suitable for type 2 diabetes research
  • For research use only
Catalog No. 50-003-44048 Supplier Medchemexpress LLC Supplier No. HY11019710MM/1ML
6bK TFA is a potent and selective inhibitor of insulin degrading enzyme (IDE) with an IC50 value of 50 nM. It increases circulating insulin in high-fat-fed mice and enhances glucose tolerance to oral glucose, notably to a greater extent in high-fat-fed mice. This product is suitable for type 2 diabetes research.
  • Potent and selective inhibitor of insulin degrading enzyme (IDE)
  • Increases circulating insulin in high-fat-fed mice
  • Enhances glucose tolerance to oral glucose
  • Suitable for type 2 diabetes research
  • For research use only