Learn More
Medchemexpress LLC 6bK TFA | 1774353-12-6 | 99.7% | 871.94 | 25 MG
SDP
Catalog No. 5000345845
Encompass_Preferred
6bK TFA is a potent and selective insulin degrading enzyme (IDE) inhibitor with an IC50 value of 50 nM. It increases circulating insulin in high-fat-fed mice and enhances glucose tolerance to oral glucose, especially in high-fat-fed mice.
  • Potent and selective insulin degrading enzyme (IDE) inhibitor
  • IC50 value of 50 nM
  • Increases circulating insulin in high-fat-fed mice
  • Enhances glucose tolerance to oral glucose
  • Suitable for type 2 diabetes research
Catalog No. 50-003-45845 Supplier Medchemexpress LLC Supplier No. 25MGHY110197
6bK TFA is a potent and selective insulin degrading enzyme (IDE) inhibitor with an IC50 value of 50 nM. It increases circulating insulin in high-fat-fed mice and enhances glucose tolerance to oral glucose, especially in high-fat-fed mice.
  • Potent and selective insulin degrading enzyme (IDE) inhibitor
  • IC50 value of 50 nM
  • Increases circulating insulin in high-fat-fed mice
  • Enhances glucose tolerance to oral glucose
  • Suitable for type 2 diabetes research