Learn More
Medchemexpress LLC 6bK (TFA) | 1774353-12-6 | MFCD30182289 | 99.4% | 871.94 g/mol | C43H56F3N7O9 | 5 MG
SDP
Catalog No. 5000264202
Encompass_Preferred
6bK TFA is a potent, selective insulin-degrading enzyme (IDE) inhibitor used in preclinical research to study insulin metabolism and glucose tolerance. It shows an in vitro IC50 of approximately 50 nM and has been reported to increase circulating insulin and improve oral glucose tolerance in high-fat-fed mice.
  • Potent IDE inhibition (IC50 ≈ 50 nM).
  • Selective activity for insulin-degrading enzyme.
  • High purity (≈99.4%).
  • Available as solid and DMSO solution in multiple pack sizes.
  • Supplied with analytical documents (COA, HPLC, LCMS).
  • Recommended storage: solid -20°C; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-64202 Supplier Medchemexpress LLC Supplier No. HY1101975MG
6bK TFA is a potent, selective insulin-degrading enzyme (IDE) inhibitor used in preclinical research to study insulin metabolism and glucose tolerance. It shows an in vitro IC50 of approximately 50 nM and has been reported to increase circulating insulin and improve oral glucose tolerance in high-fat-fed mice.
  • Potent IDE inhibition (IC50 ≈ 50 nM).
  • Selective activity for insulin-degrading enzyme.
  • High purity (≈99.4%).
  • Available as solid and DMSO solution in multiple pack sizes.
  • Supplied with analytical documents (COA, HPLC, LCMS).
  • Recommended storage: solid -20°C; in solvent -80°C (6 months) or -20°C (1 month).