Learn More
Cayman Chemical 6-Carboxytetramethylrhodamine, Purity: ≥95%, CAS: 91809-67-5, 50 mg
SDP
Catalog No. 502937495
Encompass_Preferred
A fluorescent dye; ex/em = 543/572 nm, respectively; has commonly been used for the covalent labeling of oligonucleotides for DNA analysis; has been used in various DNA-protein binding studies, DNA FRET experiments, and as a standard reporter or quencher dye in RT-PCR
Catalog No. 50-293-7495 Supplier Cayman Chemical Supplier No. 2904650 MG
A fluorescent dye; ex/em = 543/572 nm, respectively; has commonly been used for the covalent labeling of oligonucleotides for DNA analysis; has been used in various DNA-protein binding studies, DNA FRET experiments, and as a standard reporter or quencher dye in RT-PCR