Learn More
Cayman Chemical 6-Chloro-3-indolyl-β-D-Glucuronide (cyclohexylammonium salt), Purity: ≥95%, CAS: 138182-20-4, 100 mg
SDP
Catalog No. 502988618
Encompass_Preferred
A β-glucuronidase chromogenic substrate; upon cleavage by β-glucuronidase, an insoluble salmon precipitate is formed that can be used in histochemical analysis of β-glucuronidase activity
Catalog No. 50-298-8618 Supplier Cayman Chemical Supplier No. 30131100 MG
A β-glucuronidase chromogenic substrate; upon cleavage by β-glucuronidase, an insoluble salmon precipitate is formed that can be used in histochemical analysis of β-glucuronidase activity