Learn More
Cayman Chemical 6dIazo5oxoLnorLeucIn 10mg
SDP
Catalog No. 502965669
Encompass_Preferred
A glutamine analog that inhibits glutaminases (Ki = 6 µM) and other glutamine-utilizing enzymes, including cytidine triphosphate synthase
Catalog No. 50-296-5669 Supplier Cayman Chemical Supplier No. 1758010 MG
A glutamine analog that inhibits glutaminases (Ki = 6 µM) and other glutamine-utilizing enzymes, including cytidine triphosphate synthase