Learn More
Cayman Chemical 6-diazo-5-oxo-L-nor-Leucine, Purity: ≥95%, CAS: 157-03-9, 50 mg
SDP
Catalog No. 502966404
Encompass_Preferred
A glutamine analog that inhibits glutaminases (Ki = 6 µM) and other glutamine-utilizing enzymes, including cytidine triphosphate synthase
Catalog No. 50-296-6404 Supplier Cayman Chemical Supplier No. 1758050 MG
A glutamine analog that inhibits glutaminases (Ki = 6 µM) and other glutamine-utilizing enzymes, including cytidine triphosphate synthase