Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (6E)-3-bromo-4-(4-phenyl-1,2,3,4-tetrahydronaphthalen-1-ylidene)furan-2(5H)-one | 88070-98-8 | MFCD00270871 | 99.6% | 317.18 g/mol | C16H13BrO2 | 100 MG
SDP

Catalog No. 5000241404
Encompass_Preferred

Bromoenol lactone is an irreversible, suicide-based inhibitor of phospholipase A2 used as a research reagent to study lipid signaling, inflammation, and enzyme function. It is supplied as a white-to-yellow solid of high purity and is typically prepared in DMSO for in vitro and in vivo experiments. Powder storage at -20°C is recommended; solutions have limited stability.

  • Irreversible, suicide-based inhibitor of phospholipase A2.
  • High purity suitable for biochemical and pharmacological assays (≈99.6%).
  • Molecular weight 317.18 g/mol; chemical formula C16H13BrO2.
  • Soluble in DMSO (≈50 mg/mL); sonication recommended for dissolution.
  • Powder stability at -20°C for up to 3 years; limited in-solution shelf life.
  • Supplied as a solid to allow flexible formulation for in vitro and in vivo use.

Catalog No. 50-002-41404 Supplier Medchemexpress LLC Supplier No. HY107411100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoenol lactone is an irreversible, suicide-based inhibitor of phospholipase A2 used as a research reagent to study lipid signaling, inflammation, and enzyme function. It is supplied as a white-to-yellow solid of high purity and is typically prepared in DMSO for in vitro and in vivo experiments. Powder storage at -20°C is recommended; solutions have limited stability.

  • Irreversible, suicide-based inhibitor of phospholipase A2.
  • High purity suitable for biochemical and pharmacological assays (≈99.6%).
  • Molecular weight 317.18 g/mol; chemical formula C16H13BrO2.
  • Soluble in DMSO (≈50 mg/mL); sonication recommended for dissolution.
  • Powder stability at -20°C for up to 3 years; limited in-solution shelf life.
  • Supplied as a solid to allow flexible formulation for in vitro and in vivo use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.