Learn More
Perkin Elmer US LLC Standard PVC Three-Stop Peristaltic Pump Tubing - 1.14 mm I.D., Red/Red/Red, Pkg. 12
Catalog No. 509051991
Encompass_Preferred
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. This clear tubing ships as a package of 12.
Catalog No. 50-905-1991 Supplier Perkin Elmer US LLC Supplier No. B0193160
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. This clear tubing ships as a package of 12.