Learn More
Aat Bioquest 6-FAM Alkyne
SDP
Catalog No. 502979210
Encompass_Preferred
6-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.
Catalog No. 50-297-9210 Supplier Aat Bioquest Supplier No. 134
6-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.