Learn More
Aat Bioquest 6-FAM, SE [6-Carboxyfluorescein, succinimidyl ester] [CAS 92557-81-8]
SDP
Catalog No. 502979209
Encompass_Preferred
6-FAM, SE is the amine-reactive succinimidyl ester of single isomer 6-FAM acid.
Catalog No. 50-297-9209 Supplier Aat Bioquest Supplier No. 118
6-FAM, SE is the amine-reactive succinimidyl ester of single isomer 6-FAM acid.