Learn More
Cayman Chemical 6-Formylindolo[3,2-b]carbazole, Purity: ≥95%, CAS: 172922-91-7, 10 mg
SDP
Catalog No. 502984447
Encompass_Preferred
A degradation product of tryptophan upon exposure to visible light that activates the AhR (Kd = 70 pM) and is a substrate for CYP1A1, CYP1A2, and CYP1B1 (Kis = 15.4, 3.9, and 13.3 µM, respectively); used to study the immunomodulatory and host defense mechanisms of AhR signaling pathways
Catalog No. 50-298-4447 Supplier Cayman Chemical Supplier No. 1952910 MG
A degradation product of tryptophan upon exposure to visible light that activates the AhR (Kd = 70 pM) and is a substrate for CYP1A1, CYP1A2, and CYP1B1 (Kis = 15.4, 3.9, and 13.3 µM, respectively); used to study the immunomodulatory and host defense mechanisms of AhR signaling pathways