Learn More
Air Science Usa Laminar Flow Cabinet 72" Wide
SDP
Catalog No. NC3735237
Encompass_Preferred
The Purair Lf Series is a 6 ft vertical laminar flow cabinet designed to provide a sterile workspace for non-hazardous materials. Its advanced ULPA filtration captures 99.999% of particles while the durable stainless steel interior ensures longevity and chemical resistance. Ideal for environments where contamination must be minimized, it features user-friendly controls and operates on a 115V electrical supply, distinguishing it from standard models with superior protection and monitoring capabilities
Catalog No. NC3735237 Supplier Air Science Usa Supplier No. VLF72SSA
The Purair Lf Series is a 6 ft vertical laminar flow cabinet designed to provide a sterile workspace for non-hazardous materials. Its advanced ULPA filtration captures 99.999% of particles while the durable stainless steel interior ensures longevity and chemical resistance. Ideal for environments where contamination must be minimized, it features user-friendly controls and operates on a 115V electrical supply, distinguishing it from standard models with superior protection and monitoring capabilities