Learn More
Medchemexpress LLC 6H-dibenzo[b,d]pyran-6-one, 8-(1-hydroxyethyl)-2-methoxy-3-[(4-methoxyphenyl)methoxy]- | 914913-88-5 | MFCD18633224 | 99.3% | C24H22O6 | 10MG
SDP
Catalog No. 5000261568
Encompass_Preferred
Palomid 529 is a small-molecule research inhibitor that targets both mTOR complex 1 (mTORC1) and mTOR complex 2 (mTORC2), modulating the PI3K/Akt/mTOR signaling pathway. It has been used in preclinical studies and early clinical research to reduce tumor growth, angiogenesis, and vascular permeability, and has been investigated for neovascular age-related macular degeneration.
  • Dual inhibition of mTORC1 and mTORC2.
  • Documented in peer-reviewed literature and public chemical databases.
  • High reported purity suitable for analytical use.
  • Suitable as an analytical standard and research reagent.
  • Provided with batch-specific certificate of analysis data.
Catalog No. 50-002-61568 Supplier Medchemexpress LLC Supplier No. HY1458110MG
Palomid 529 is a small-molecule research inhibitor that targets both mTOR complex 1 (mTORC1) and mTOR complex 2 (mTORC2), modulating the PI3K/Akt/mTOR signaling pathway. It has been used in preclinical studies and early clinical research to reduce tumor growth, angiogenesis, and vascular permeability, and has been investigated for neovascular age-related macular degeneration.
  • Dual inhibition of mTORC1 and mTORC2.
  • Documented in peer-reviewed literature and public chemical databases.
  • High reported purity suitable for analytical use.
  • Suitable as an analytical standard and research reagent.
  • Provided with batch-specific certificate of analysis data.