Learn More
Medchemexpress LLC 6H-Purin-6-one, 2-amino-1,9-dihydro-9-β-L-ribofuranosyl- | 26578-09-6 | 98.0% | 283.24 | 250 MG
SDP
Catalog No. 5000350159
Encompass_Preferred
L-Guanosine is the L-configuration of Guanosine. This purine nucleoside exhibits anti-herpesvirus activity and can be used in the preparation of supramolecular hydrogel.
  • L-configuration of guanosine
  • Purine nucleoside
  • Exhibits anti-herpesvirus activity
  • Can be used in the preparation of supramolecular hydrogel
  • Functions as a nucleoside antimetabolite/analog
  • Acts as an inhibitor
Catalog No. 50-003-50159 Supplier Medchemexpress LLC Supplier No. 250MGHY138130
L-Guanosine is the L-configuration of Guanosine. This purine nucleoside exhibits anti-herpesvirus activity and can be used in the preparation of supramolecular hydrogel.
  • L-configuration of guanosine
  • Purine nucleoside
  • Exhibits anti-herpesvirus activity
  • Can be used in the preparation of supramolecular hydrogel
  • Functions as a nucleoside antimetabolite/analog
  • Acts as an inhibitor