Learn More
Medchemexpress LLC 6H-purin-6-one, 2-amino-1,9-dihydro-9-β-l-ribofuranosyl- | 26578-09-6 | 98.0% | 283.24 | 50 MG
SDP
Catalog No. 5000348595
Encompass_Preferred
L-Guanosine is the L-configuration of Guanosine, a purine nucleoside with anti-herpesvirus activity. It can be utilized in the preparation of supramolecular hydrogel.
  • L-configuration of guanosine
  • Purine nucleoside with anti-herpesvirus activity
  • Utilized in the preparation of supramolecular hydrogel
Catalog No. 50-003-48595 Supplier Medchemexpress LLC Supplier No. HY13813050MG
L-Guanosine is the L-configuration of Guanosine, a purine nucleoside with anti-herpesvirus activity. It can be utilized in the preparation of supramolecular hydrogel.
  • L-configuration of guanosine
  • Purine nucleoside with anti-herpesvirus activity
  • Utilized in the preparation of supramolecular hydrogel