Learn More
Medchemexpress LLC 6H-Purin-6-one, 2-amino-9-[(Z)-[2,2-bis(hydroxymethyl)cyclopropylidene]methyl]-1,9-dihydro- | 632325-71-4 | 98.0% | 263.26 | 5 MG
SDP
Catalog No. 5000393889
Encompass_Preferred
Cyclopropavir (Filociclovir) is an orally active, broad-spectrum anti-herpesvirus compound that inhibits UL97 kinase activity. It exhibits antiviral activity against HCMV, MCMV, and HAdV6 viruses.
  • Orally active.
  • Broad-spectrum anti-herpesvirus compound.
  • Inhibits UL97 kinase activity.
  • Antiviral activity against HCMV, MCMV, and HAdV6 viruses.
Catalog No. 50-003-93889 Supplier Medchemexpress LLC Supplier No. HY167215MG
Cyclopropavir (Filociclovir) is an orally active, broad-spectrum anti-herpesvirus compound that inhibits UL97 kinase activity. It exhibits antiviral activity against HCMV, MCMV, and HAdV6 viruses.
  • Orally active.
  • Broad-spectrum anti-herpesvirus compound.
  • Inhibits UL97 kinase activity.
  • Antiviral activity against HCMV, MCMV, and HAdV6 viruses.