Learn More
Medchemexpress LLC 6H-purin-6-one-8-13C, 2-amino-1,7-dihydro | 201489-20-5 | 99.5% | 152.12 | C4(13C)H5N5O | 10mg
SDP
Catalog No. 5000267365
Encompass_Preferred
Guanine-13C is the 13C labeled Guanine[1] Guanine (2-Aminohypoxanthine) is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds
Catalog No. 50-002-67365 Supplier Medchemexpress LLC Supplier No. HYY1055S10MG
Guanine-13C is the 13C labeled Guanine[1] Guanine (2-Aminohypoxanthine) is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds