Learn More
Medchemexpress LLC 6H-Purine-6-thione, 1,2,3,9-tetrahydro-, hydrate (1:1) | 6112-76-1 | 99.9% | 170.19 | 500 MG
SDP
Catalog No. 5000348220
Encompass_Preferred
6-Mercaptopurine hydrate is a purine analogue that acts as an antagonist of endogenous purines. It has been widely used as an antileukemic agent and immunosuppressive agent.
  • Induces NR4A3 transcriptional activity.
  • Increases NR4A3 protein levels.
  • Increases cell surface GLUT4 in basal and insulin-stimulated cells.
  • Increases phospho-AS160.
  • Significantly inhibits EcSTH-induced phosphorylation of AMPK and ACC1.
  • Promotes an increase in S phase cell population in fetal telencephalons.
  • Promotes an increase in G2/M phase cell population in fetal telencephalons.
  • Increases sub-G1 phase cell population (apoptotic cells).
  • Inhibited expression of CAV1 protein in MM1S cells.
  • Further promoted bortezomib-mediated survival of MM-bearing mice.
Catalog No. 50-003-48220 Supplier Medchemexpress LLC Supplier No. HY13677A500MG
6-Mercaptopurine hydrate is a purine analogue that acts as an antagonist of endogenous purines. It has been widely used as an antileukemic agent and immunosuppressive agent.
  • Induces NR4A3 transcriptional activity.
  • Increases NR4A3 protein levels.
  • Increases cell surface GLUT4 in basal and insulin-stimulated cells.
  • Increases phospho-AS160.
  • Significantly inhibits EcSTH-induced phosphorylation of AMPK and ACC1.
  • Promotes an increase in S phase cell population in fetal telencephalons.
  • Promotes an increase in G2/M phase cell population in fetal telencephalons.
  • Increases sub-G1 phase cell population (apoptotic cells).
  • Inhibited expression of CAV1 protein in MM1S cells.
  • Further promoted bortezomib-mediated survival of MM-bearing mice.