Learn More
Medchemexpress LLC 6H-Thieno[3,2-f]triazolo[4,3-a]diazepine-6-acetamide, 4-(4-chlorophenyl)-N-[6-[[[2-(2,6-dioxo | 1957234-97-7 | 99.4% | 783.29 | 10 MG
SDP
Catalog No. 5000437776
Encompass_Preferred
PROTAC BET Degrader-10 (dBET37) is a potent BET protein BRD4 degrader, connected by ligands for Cereblon and BRD4, with a DC50 of 49 nM. This compound contains two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • Potent BET protein BRD4 degrader
  • Utilizes the intracellular ubiquitin-proteasome system for selective degradation
  • Useful for research applications
Catalog No. 50-004-37776 Supplier Medchemexpress LLC Supplier No. HY11271810MG
PROTAC BET Degrader-10 (dBET37) is a potent BET protein BRD4 degrader, connected by ligands for Cereblon and BRD4, with a DC50 of 49 nM. This compound contains two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • Potent BET protein BRD4 degrader
  • Utilizes the intracellular ubiquitin-proteasome system for selective degradation
  • Useful for research applications