Learn More
Medchemexpress LLC 6H-thieno[3,2-f]triazolo[4,3-a]diazepine-6-acetamide, 4-(4-chlorophenyl)-N-[6-[[[2-(2 ,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]methyl]amino]-6-oxohexyl]-2,3,9- trimethyl | 1957234-97-7 | 99.4% | 783.29 | 100 MG
SDP
Catalog No. 5000437671
Encompass_Preferred
PROTAC BET Degrader-10 (dBET37) is a potent degrader of the BET protein BRD4, featuring a connection between ligands for Cereblon and BRD4, with a DC50 of 49 nM. The composition includes a BRD4(BD1BD2) ligand, a Cereblon ligand, and a linker. This product is intended for research use only.
  • It is a potent BET protein BRD4 degrader.
  • It is connected by ligands for Cereblon and BRD4.
  • It has a DC50 of 49 nM.
  • Purity is 99.39%.
  • It is available in solid form.
  • The color is off-white to light yellow.
  • It can be shipped at room temperature in the continental US.
  • Powder storage conditions are -20°C for 3 years or 4°C for 2 years.
  • In solvent, storage conditions are -80°C for 6 months or -20°C for 1 month.
  • It is soluble in DMSO at 100 mg/mL (127.67 mM).
Catalog No. 50-004-37671 Supplier Medchemexpress LLC Supplier No. HY112718100MG
PROTAC BET Degrader-10 (dBET37) is a potent degrader of the BET protein BRD4, featuring a connection between ligands for Cereblon and BRD4, with a DC50 of 49 nM. The composition includes a BRD4(BD1BD2) ligand, a Cereblon ligand, and a linker. This product is intended for research use only.
  • It is a potent BET protein BRD4 degrader.
  • It is connected by ligands for Cereblon and BRD4.
  • It has a DC50 of 49 nM.
  • Purity is 99.39%.
  • It is available in solid form.
  • The color is off-white to light yellow.
  • It can be shipped at room temperature in the continental US.
  • Powder storage conditions are -20°C for 3 years or 4°C for 2 years.
  • In solvent, storage conditions are -80°C for 6 months or -20°C for 1 month.
  • It is soluble in DMSO at 100 mg/mL (127.67 mM).