Learn More
Medchemexpress LLC 6H-Thieno[3,2-f]triazolo[4,3-a]diazepine-6-acetic acid, 4-(4-chlorophenyl)-2,3,9-trimethyl-, cyclopropyl ester, (6S)- | 3077900-08-1 | 98.3% | 440.95 | 25 MG
SDP
Catalog No. 5000384892
Encompass_Preferred
DDL-357 is a potent secreted clusterin enhancer. It reduces phospho-tau in the brain and improves memory in the murine 3xTg-AD model. It is for research use only.
  • Potent secreted clusterin enhancer.
  • Reduces phospho-tau in the brain.
  • Improves memory in the murine 3xTg-AD model.
Catalog No. 50-003-84892 Supplier Medchemexpress LLC Supplier No. 25MGHY176438
DDL-357 is a potent secreted clusterin enhancer. It reduces phospho-tau in the brain and improves memory in the murine 3xTg-AD model. It is for research use only.
  • Potent secreted clusterin enhancer.
  • Reduces phospho-tau in the brain.
  • Improves memory in the murine 3xTg-AD model.