Learn More
TARGETMOL CHEMICALS INC 6H05 5MG
SDP
Catalog No. 502577270
Encompass_Preferred
Also available in 10mg 25mg 50mg and bulk. Please contact Fisher for quotes. 6H05 (K-Ras inhibitor) is a selective allosteric inhibitor of oncogenic mutant K-Ras(G12C). purity: 98%
Catalog No. 50-257-7270 Supplier TARGETMOL CHEMICALS INC Supplier No. T19315MG
Also available in 10mg 25mg 50mg and bulk. Please contact Fisher for quotes. 6H05 (K-Ras inhibitor) is a selective allosteric inhibitor of oncogenic mutant K-Ras(G12C). purity: 98%