Learn More
TARGETMOL CHEMICALS INC 6H05 TFA 5MG
SDP
Catalog No. 502576327
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. 6H05 TFA (6H05 trifluoroacetate) is a selective and allosteric oncogenic mutant K-Ras(G12C) inhibitor. purity: 98%
Catalog No. 50-257-6327 Supplier TARGETMOL CHEMICALS INC Supplier No. T73265MG
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. 6H05 TFA (6H05 trifluoroacetate) is a selective and allosteric oncogenic mutant K-Ras(G12C) inhibitor. purity: 98%