Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6H05 trifluoroacetate | 2061344-88-3 | 99.0% | 590.14 | C22H31ClF3N3O4S3 | 1MG
SDP

Catalog No. 5000236026
Encompass_Preferred

6H05 TFA is the trifluoroacetate salt of 6H05, a selective, allosteric inhibitor of the oncogenic mutant K-Ras(G12C) intended for biochemical and cellular research. Supplied as a solid or as a 10 mM DMSO solution, the compound is provided at high purity for assay development and mechanistic studies.

  • Selective, allosteric inhibitor of K-Ras(G12C).
  • High purity (99.02%).
  • Molecular formula C22H31ClF3N3O4S3; molecular weight 590.14.
  • Available in small-scale solid quantities and as a 10 mM DMSO solution.
  • Suitable for biochemical and cellular assays; for research use only, not for human use.

Catalog No. 50-002-36026 Supplier Medchemexpress LLC Supplier No. HY12408A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6H05 TFA is the trifluoroacetate salt of 6H05, a selective, allosteric inhibitor of the oncogenic mutant K-Ras(G12C) intended for biochemical and cellular research. Supplied as a solid or as a 10 mM DMSO solution, the compound is provided at high purity for assay development and mechanistic studies.

  • Selective, allosteric inhibitor of K-Ras(G12C).
  • High purity (99.02%).
  • Molecular formula C22H31ClF3N3O4S3; molecular weight 590.14.
  • Available in small-scale solid quantities and as a 10 mM DMSO solution.
  • Suitable for biochemical and cellular assays; for research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.