Learn More
Biomatik Corporation 6*His Monoclonal Antibody
Supplier:  Biomatik Corporation CAC10284
Encompass_Preferred
6*His Monoclonal Antibody is a Monoclonal Antibody to 6*His. The 6*His Monoclonal Antibody has been validated for the following applications: ELISA, WB. Host: Mouse. Immunogen: Hhhhhh (6X His) Synthetic Peptide Conjugate To Klh. Applications: ELISA, WB. Target Synonyms: 6*His. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6350