Learn More
Sklar Instruments Forceps, Gillies Forceps, Sklar 6 in., 1x2 Teeth, Straight, Cross-Serrated Jaws, Stainless Steel, EA.
SDP
Catalog No. 501098030 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Gillies Forceps, Sklar, 6 in., 1x2 Teeth, Straight, Cross-Serrated Jaws, Stainless Steel, EA. Sklar Gillies Forceps are thumb forceps often used in small surgical sites to grasp tissue. They have fine, narrow jaws that are commonly used in plastic surgery. This product is straight with cross-serrated tips, 1x2 teeth, and a length of 6 inches.
Catalog No. 50-109-8030 Supplier Sklar Instruments Supplier No. 192070
Forceps, Gillies Forceps, Sklar, 6 in., 1x2 Teeth, Straight, Cross-Serrated Jaws, Stainless Steel, EA. Sklar Gillies Forceps are thumb forceps often used in small surgical sites to grasp tissue. They have fine, narrow jaws that are commonly used in plastic surgery. This product is straight with cross-serrated tips, 1x2 teeth, and a length of 6 inches.