Learn More
Sklar Instruments Forceps, Semken Dressing Forceps, Sklar 6 in., Curved, Serrated, Narrow Tip, Thumb, Stainless Steel, EA.
SDP
Catalog No. 501098143 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Semken Dressing Forceps, Sklar, 6 in., Curved, Serrated, Narrow Tip, Thumb, Stainless Steel, EA. Sklar Semken Dressing Forceps are most commonly used for application and changing of dressings. These forceps feature narrow tips that may be beneficial for neurosurgery or other procedures performed in constricted areas. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. This product is curved with serrated tips and a length of 6 inches.
Catalog No. 50-109-8143 Supplier Sklar Instruments Supplier No. 224156
Forceps, Semken Dressing Forceps, Sklar, 6 in., Curved, Serrated, Narrow Tip, Thumb, Stainless Steel, EA. Sklar Semken Dressing Forceps are most commonly used for application and changing of dressings. These forceps feature narrow tips that may be beneficial for neurosurgery or other procedures performed in constricted areas. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. This product is curved with serrated tips and a length of 6 inches.