Learn More
Sklar Instruments Scissors, Sklar Tungsten Carbide Supercut Sklar Mayo-Stille Dissecting Scissors, Sklar 6 in., Blunt/Blunt, Straight, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Tungsten Carbide / Stainless Steel, EA.
SDP
Catalog No. 501093573 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sklar Edge Tungsten Carbide Supercut Sklarhone Mayo-Stille Dissecting Scissors, Sklar, 6 in., Blunt/Blunt, Straight, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Tungsten Carbide / Stainless Steel, EA. Sklar Edge Tungsten Carbide Supercut Sklarhone Mayo-Stille Dissecting Scissors provide effortless, precise cutting with the benefit of the strength and durability of tungsten carbide. These scissors feature one knife-like razor edge and one micro-serrated scissors edge and can be used to cut both delicate and heavy tissue with minimal tearing or pulling. Sklar Edge Tungsten Carbide Scissors provide an exceptionally hard and durable cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than other non tungsten carbide scissors.
Catalog No. 50-109-3573 Supplier Sklar Instruments Supplier No. 163572
Scissors, Sklar Edge Tungsten Carbide Supercut Sklarhone Mayo-Stille Dissecting Scissors, Sklar, 6 in., Blunt/Blunt, Straight, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Tungsten Carbide / Stainless Steel, EA. Sklar Edge Tungsten Carbide Supercut Sklarhone Mayo-Stille Dissecting Scissors provide effortless, precise cutting with the benefit of the strength and durability of tungsten carbide. These scissors feature one knife-like razor edge and one micro-serrated scissors edge and can be used to cut both delicate and heavy tissue with minimal tearing or pulling. Sklar Edge Tungsten Carbide Scissors provide an exceptionally hard and durable cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than other non tungsten carbide scissors.