Learn More
SAI Infusion Technologies 10 Gauge 6" Trocar - Semi-Blunt 45° Beveled Tip
SDP
Catalog No. NC0242308
Encompass
6" Trocar for Rodent Surgery - 10 Gauge, with a semi-blunt 45° Beveled Tip. Subcutaneous tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes under the skin. SAI's TRO-10-6 is manufactured from thick-walled medical-grade stainless steel, has a semi-blunt tip and is ideal for rat surgery. Customized tips and lengths are available, please contact us for more information. 10G, 6 in. Product is for laboratory animal research only.
Catalog No. NC0242308 Supplier SAI Infusion Technologies Supplier No. TRO106
6" Trocar for Rodent Surgery - 10 Gauge, with a semi-blunt 45° Beveled Tip. Subcutaneous tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes under the skin. SAI's TRO-10-6 is manufactured from thick-walled medical-grade stainless steel, has a semi-blunt tip and is ideal for rat surgery. Customized tips and lengths are available, please contact us for more information. 10G, 6 in. Product is for laboratory animal research only.