Learn More
Ace Glass, Inc. 6L jacketed filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx350mm depth, with CAPFE O-ring, #15 Ace-Thred inlet/outlet connections, use with 1/2in ID tubing, pressure rated at 35 psi.
SDP
Catalog No. 50972811
Encompass
6L jacketed filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx350mm depth, with CAPFE O-ring, #15 Ace-Thred inlet/outlet connections, use with 1/2in ID tubing, pressure rated at 35 psi.
Catalog No. 50-972-811 Supplier Ace Glass, Inc. Supplier No. 6386328
6L jacketed filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx350mm depth, with CAPFE O-ring, #15 Ace-Thred inlet/outlet connections, use with 1/2in ID tubing, pressure rated at 35 psi.