Learn More
Chemglass Life Sciences 6L Bioreactor Vessel / 9L Carboy Carrier
SDP
Catalog No. 501532134
Encompass
Manufactured from 304 Stainless Steel and electro-polished.Vessel carriers are very stable and allow for easy transport of Bioreactor Vessels, Spinner Flasks or large Carboys.Dimensions/H x OAD (in.): CLS-1406-C06: 9 x 8 CLS-1406-C08: 11 x 10 CLS-1406-C15: 12 x 11 CLS-1406-C36: 17 x 13
Catalog No. 50-153-2134 Supplier Chemglass Life Sciences Supplier No. CLS1406C06
Manufactured from 304 Stainless Steel and electro-polished.Vessel carriers are very stable and allow for easy transport of Bioreactor Vessels, Spinner Flasks or large Carboys.Dimensions/H x OAD (in.): CLS-1406-C06: 9 x 8 CLS-1406-C08: 11 x 10 CLS-1406-C15: 12 x 11 CLS-1406-C36: 17 x 13