Learn More
Ace Glass, Inc. Mantle, 6L, for 235mm diameter spherical flask, 235mm ID x 120.6mm height, 700w-115v, CSA rated
SDP
Catalog No. 50977326
Encompass
6L aluminum housed, table-model mantle for 235mm diameter spherical flask. 235mm I.D. x 120.6mm height. Maximum internal operating temperature is 450 Deg. centigrade. 700w-115v, cULus rated. Power cord included.
Catalog No. 50-977-326 Supplier Ace Glass, Inc. Supplier No. 1205326
6L aluminum housed, table-model mantle for 235mm diameter spherical flask. 235mm I.D. x 120.6mm height. Maximum internal operating temperature is 450 Deg. centigrade. 700w-115v, cULus rated. Power cord included.