Learn More
Chemglass Life Sciences 6000mL Erlenmeyer Flask, 38-430 GPI Screw Thread
SDP
Catalog No. CG154309
Encompass_Preferred
6000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. CG154309 Supplier Chemglass Life Sciences Supplier No. CG154309
6000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.