Learn More
Cayman Chemical 6methoxy Naphthalen ACtIc 50mg
SDP
Catalog No. 502895186
Encompass_Preferred
6-MNA is a competitive, non-selective COX inhibitor.{1286,1364,1791} The Ki values for ovine COX-1 and -2 are 21 and 19 µM, respectively.{1364} The IC50 values are 70 and 20 µM for human recombinant COX-1 and -2, respectively.{1286}
Catalog No. 50-289-5186 Supplier Cayman Chemical Supplier No. 7062050 MG
6-MNA is a competitive, non-selective COX inhibitor.{1286,1364,1791} The Ki values for ovine COX-1 and -2 are 21 and 19 µM, respectively.{1364} The IC50 values are 70 and 20 µM for human recombinant COX-1 and -2, respectively.{1286}