Learn More
MSC P/F DISPOSABLE GLOVES
Catalog No. 502739165
Encompass
Disposable/Single Use Gloves; Primary Material: Nitrile ; Thickness (mil): 7.5 ; Grip Surface: Textured (Fingertips) ; Grade: Food ; Cuff Type: Extended ; Length (Inch): 11.8
Catalog No. 50-273-9165 Supplier MSC Supplier No. 08717555
Disposable/Single Use Gloves; Primary Material: Nitrile ; Thickness (mil): 7.5 ; Grip Surface: Textured (Fingertips) ; Grade: Food ; Cuff Type: Extended ; Length (Inch): 11.8