Learn More
WELCH MATERIALS INC 6mL Clear Glass 22×38mm Crimp Headspace Vial. 20mm Beveled Edge. Flat Bottom. Short Neck. 100pcs/pk.
Catalog No. NC3594597
Encompass_Preferred
Brand: Doprah, Vial Type: Headspace Vial, Closure Type: Crimp, Volume Capacity: 6mL, Dimension: 22×38mm, Vial Material: Clear Glass
Catalog No. NC3594597 Supplier WELCH MATERIALS INC Supplier No. DE1132
Brand: Doprah, Vial Type: Headspace Vial, Closure Type: Crimp, Volume Capacity: 6mL, Dimension: 22×38mm, Vial Material: Clear Glass