Learn More
Alkali Scientific 6mL Glass Crimp Top Headspace
SDP
Catalog No. 503027031
Encompass_Preferred
Designed for high-performance chromatography analysis ASI6 mL Clear Glass Flat-Bottom Crimp Top Headspace Vials offer an affordable high-quality solution compatible with all headspace GC instruments Manufactured from Type I borosilicate glass these vials provide exceptional thermal and chemical resistance making them ideal for volatile gas analysis The flat-bottom design improves heating efficiency and is well-suited for manual headspace autosamplers Key Features Superior Glass Quality Made from 1st hydrolytic class borosilicate glass (Type I) for enhanced chemical resistance and durability Beveled Finish Reduces the sealing surface between the crimp seal and the glass for a more secure and consistent closure Standard 20 mm Crimp Top Ensures compatibility with a wide range of crimp seals and autosamplers Optimal Size for Headspace Analysis 22 5 32 mm dimensions allow precise sample containment Flat Bottom Design Preferred for manual headspace autosamplers and applications
Catalog No. 50-302-7031 Supplier Alkali Scientific Supplier No. GC006FB
Designed for high-performance chromatography analysis ASI6 mL Clear Glass Flat-Bottom Crimp Top Headspace Vials offer an affordable high-quality solution compatible with all headspace GC instruments Manufactured from Type I borosilicate glass these vials provide exceptional thermal and chemical resistance making them ideal for volatile gas analysis The flat-bottom design improves heating efficiency and is well-suited for manual headspace autosamplers Key Features Superior Glass Quality Made from 1st hydrolytic class borosilicate glass (Type I) for enhanced chemical resistance and durability Beveled Finish Reduces the sealing surface between the crimp seal and the glass for a more secure and consistent closure Standard 20 mm Crimp Top Ensures compatibility with a wide range of crimp seals and autosamplers Optimal Size for Headspace Analysis 22 5 32 mm dimensions allow precise sample containment Flat Bottom Design Preferred for manual headspace autosamplers and applications