Learn More
Research Products International Corp 6ml High Density Polyethylene Vial, Push-On Caps, Bulk Packed, 2000 per Case
SDP
Catalog No. 50998445
Encompass
This general purpose vial can be used for liquid scintillation counting, gamma counting, chromatography, sample storage and culturing. The shoulderless vial features a full width opening of 12.5 mm and the overall dimensions are 16 x 57 mm.
The vials can be securely sealed by simply snapping the caps on; removal of caps requires an easy 1/4 turn (twist). The ease with which these caps can be manipulated eliminates the danger of spillage associated with other push-on/pull-off caps. A built-in positive lock prevents the cap from popping off because of a small build-up of pressure in the vial. The quality of the sealing system is such that it is not necessary to tighten the closure with pressure to achieve a leak proof seal.
Catalog No. 50-998-445 Supplier Research Products International Corp Supplier No. 125514
This general purpose vial can be used for liquid scintillation counting, gamma counting, chromatography, sample storage and culturing. The shoulderless vial features a full width opening of 12.5 mm and the overall dimensions are 16 x 57 mm.
The vials can be securely sealed by simply snapping the caps on; removal of caps requires an easy 1/4 turn (twist). The ease with which these caps can be manipulated eliminates the danger of spillage associated with other push-on/pull-off caps. A built-in positive lock prevents the cap from popping off because of a small build-up of pressure in the vial. The quality of the sealing system is such that it is not necessary to tighten the closure with pressure to achieve a leak proof seal.