Learn More
Across International 6mm Diameter ID Vacuum Dry Pressing Die Set
SDP
Catalog No. 502251341
Encompass_Preferred
A set of 6mm diameter (ID) hardened steel dry vacuum pressing die set for pellet presses, ready for you to make 6mm diameter cylinder-shape pellets in a vacuum condition.
This item is not returnable. View return policy
Catalog No. 50-225-1341 Supplier Across International Supplier No. SDS6.V
A set of 6mm diameter (ID) hardened steel dry vacuum pressing die set for pellet presses, ready for you to make 6mm diameter cylinder-shape pellets in a vacuum condition.