Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, 6mm Bore, 20/44 Plug Size, 12mm OD Sidearms, #5 Clip Size
SDP
Catalog No. CG40005
Encompass_Preferred
Stopcock, Straight Bore, Glass Plug, 6mm Bore, 20/44 Plug Size, 12mm OD Sidearms, #5 Clip Size. The 6mm bore sizes are supplied complete with the listed size metal retaining clip CG-423.
Catalog No. CG40005 Supplier Chemglass Life Sciences Supplier No. CG40005
Stopcock, Straight Bore, Glass Plug, 6mm Bore, 20/44 Plug Size, 12mm OD Sidearms, #5 Clip Size. The 6mm bore sizes are supplied complete with the listed size metal retaining clip CG-423.