Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, 1-Arm Larger, 6mm Bore, 20/44 Plug Size, 12mm & 16mm OD Sidearms, #5 Clip Sizeer
SDP
Catalog No. CG40203
Encompass_Preferred
Stopcock, Straight Bore, Glass Plug, 1-Arm Larger, 6mm Bore, 20/44 Plug Size, 12mm & 16mm OD Sidearms, #5 Clip Sizeer. Similar to CG-400, but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Supplied complete with the listed size metal retaining clip CG-423.FOR REPLACEMENT PLUGS, SEE CG-400 LISTING
Catalog No. CG40203 Supplier Chemglass Life Sciences Supplier No. CG40203
Stopcock, Straight Bore, Glass Plug, 1-Arm Larger, 6mm Bore, 20/44 Plug Size, 12mm & 16mm OD Sidearms, #5 Clip Sizeer. Similar to CG-400, but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Supplied complete with the listed size metal retaining clip CG-423.FOR REPLACEMENT PLUGS, SEE CG-400 LISTING