Learn More
Ace Glass, Inc. 6MM GRADED SEALS 6MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845508
Encompass
6mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-979-780