Learn More
Across International 6mm ID Pressing Die Set w/ 2 Push Rods
SDP
Catalog No. 502251327
Encompass_Preferred
A set of 6mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 6mm diameter cylinder-shape pellets.
This item is not returnable. View return policy
Catalog No. 50-225-1327 Supplier Across International Supplier No. SDS6
A set of 6mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 6mm diameter cylinder-shape pellets.