Learn More
Ace Glass, Inc. Glass filled PTFE lock nut designed for 6mm OD Trubore stirring shaft bearing assembly.
SDP
Catalog No. 50979643
Encompass_Preferred
6mm glass-filled PTFE lock nut, for use with self-aligning, lubricant free PTFE Trubore bearing stir shaft assembly. Use with 6mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-979-643 Supplier Ace Glass, Inc. Supplier No. 8066107
6mm glass-filled PTFE lock nut, for use with self-aligning, lubricant free PTFE Trubore bearing stir shaft assembly. Use with 6mm O.D. stainless steel or polished glass shafts.