Learn More
Ace Glass, Inc. 6mm PTFE inner bearing, straight adapter style
SDP
Catalog No. 50975964
Encompass_Preferred
6mm self-aligning, lubricant-free PTFE inner Trubore bearing, straight adapter style. Use with ACE polished stainless steel or glass 6mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-975-964 Supplier Ace Glass, Inc. Supplier No. 8066102
6mm self-aligning, lubricant-free PTFE inner Trubore bearing, straight adapter style. Use with ACE polished stainless steel or glass 6mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.