Learn More
Chemglass Life Sciences 6mm Bore, 16/35 Plug Size, 12mm OD Sidearm
SDP
Catalog No. 501531993
Encompass_Preferred
6mm Bore, 16/35 Plug Size, 12mm OD Sidearm. Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. 50-153-1993 Supplier Chemglass Life Sciences Supplier No. CGB42407
6mm Bore, 16/35 Plug Size, 12mm OD Sidearm. Straight bore stopcock having a 1:5 taper solid PTFE plug.