Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Plug, 6mm Bore, 16/35 Plug Size, 12mm OD Sidearms
SDP
Catalog No. CG42407
Encompass_Preferred
Stopcock, Straight Bore, PTFE Plug, 6mm Bore, 16/35 Plug Size, 12mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. CG42407 Supplier Chemglass Life Sciences Supplier No. CG42407
Stopcock, Straight Bore, PTFE Plug, 6mm Bore, 16/35 Plug Size, 12mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.