Learn More
Chemglass Life Sciences Stopcock, T-Bore, PTFE Plug, 6mm Bore, 24/40 Plug Size, 12mm OD Sidearms
SDP
Catalog No. CG43603
Encompass_Preferred
Stopcock, T-Bore, PTFE Plug, 6mm Bore, 24/40 Plug Size, 12mm OD Sidearms. A 1:5 taper PTFE stopcock with three stems sealed 90º apart. PTFE plug has a "T" shaped bore to connect any two or all three stems.
Catalog No. CG43603 Supplier Chemglass Life Sciences Supplier No. CG43603
Stopcock, T-Bore, PTFE Plug, 6mm Bore, 24/40 Plug Size, 12mm OD Sidearms. A 1:5 taper PTFE stopcock with three stems sealed 90º apart. PTFE plug has a "T" shaped bore to connect any two or all three stems.